Order Entry
Germany
Orders LinkContactUsLinkComponent
Drip pads

Drip pads

Pad drip extended, 7 l, 914 × 610 mm
Catalog # 119-0349
Drip pads
Catalog # 119-0349
Drip pads
Drip pads

Pad drip extended, 7 l, 914 × 610 mm

Price
213,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Drip pads can capture small oil leaks and drips before they contaminate the ground or create unsafe work surfaces.

  • Lightweight, flexible, and portable
  • Made from UV and chemical-resistant modified PVC
  • Coated mesh outer layer to trap coarse debris

These drip pads feature replaceable, oil-only sorbent with a welded leakproof barrier. Rated for drive over applications, pads include brass grommets in opposing corners for anchoring. Anchoring stakes not included. Pads come in two sizes; extended model is weighted to keep it down in high winds. Outer shell is reusable with optional refill sorbent packs.

Certifications: Helps to comply with EPA for spill prevention (SPCC).

Specifications
  • Capacity:
    7 l
  • Description:
    Pad drip extended
  • L×W:
    914 × 610 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.