Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Plunger Dispensing Can, 1 quart, perforated pan screen serves as flame arrester, Steel, Yellow. 1 * 1 items
Catalog # 119-0261
Supplier:
JUSTRITE MANUFACTURING
Expand image
Plunger Dispensing Can, 1 quart, perforated pan screen serves as flame arrester, Steel, Yellow. 1 * 1 items
Catalog # 119-0261
Supplier:
JUSTRITE MANUFACTURING