Double spatulas, bent
150 mm, 9 mm
Catalog # USBE3292
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Stainless steel.
Specifications
- Length:150 mm
- Seq:01
- Blade width:9 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



