Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
MERCURY 1MG/L IN HNO3 5% 1 component Hg 1mg/l in HNO3 5% 1 * 100 mL
CMPLRM000967L1
:
CUSTOM MADE CHEMICALS LAB
MERCURY 1MG/L IN HNO3 5% 1 component Hg 1mg/l in HNO3 5% 1 * 100 mL
CMPLRM000967L1
:
CUSTOM MADE CHEMICALS LAB