ContactUsLinkComponent
Waterproof Pocketherm digital thermometer (thermistor) -40°C to +125°C. 1 * 1 items
Catalog # COMAP125W
Supplier:  COMARK
Waterproof Pocketherm digital thermometer (thermistor) -40°C to +125°C. 1 * 1 items
Catalog # COMAP125W
Supplier:  COMARK