ContactUsLinkComponent
RF312Glycol WiFi Datalogger + 12-months FREE Cloud Application. The Glycol Simulant Data Logger (detachable buffer probe) is designed specifically for use in monitoring the temperature of perishable items, such as Drugs, Vaccines, R&D Samples & Food. 1 * 1 items
Catalog # COMARF312GLYCOL
Supplier:  COMARK
RF312Glycol WiFi Datalogger + 12-months FREE Cloud Application. The Glycol Simulant Data Logger (detachable buffer probe) is designed specifically for use in monitoring the temperature of perishable items, such as Drugs, Vaccines, R&D Samples & Food. 1 * 1 items
Catalog # COMARF312GLYCOL
Supplier:  COMARK
Frequently Bought Together