Mop heads, polyester, quiltec I
Mop head, 390×130 mm, 5 per double bag/8 bags per case
Catalog # 568-0909
Supplier: CONTEC
Jump to:
Product Details & Documents
Mop heads are constructed from a two-ply quilted 100% polyester on the outer face with a synthetic fibrefill core.
- Designed for single use
- Multi-layer sewn lamination
- Mop heads are compatible with a wide range of chemicals
Highly sorbent mop heads are suitable for the application of disinfectants and removal of light soil, residues and biofilms.
Specifications
- Description:Mop head
- Dimensions:390×130 mm
- Seq:01
- Packed:5 per double bag/8 bags per case
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



