ContactUsLinkComponent
Human CTLA-4, His Tag (CT4-H5229) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Phe 162 (Accession NP005205.2) . 1 * 1 mg
Catalog # ACRBCT4-H5229-1MG
Supplier:  ACROBIOSYSTEMS
Human CTLA-4, His Tag (CT4-H5229) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Phe 162 (Accession NP005205.2) . 1 * 1 mg
Catalog # ACRBCT4-H5229-1MG
Supplier:  ACROBIOSYSTEMS