ContactUsLinkComponent
The horizontal laboratory autoclaves of CertoClav-Vacuum-Professional-Series are available with a chamber volume of 8 liters, 12 liters, 18 liters or 22 liters. Pre-heatable chamber, cooling-ventilation and vacuum pump guarantees fast cycles from 20 1 * 1 items
Catalog # CERT8500486
Supplier:  CERTOCLAV
The horizontal laboratory autoclaves of CertoClav-Vacuum-Professional-Series are available with a chamber volume of 8 liters, 12 liters, 18 liters or 22 liters. Pre-heatable chamber, cooling-ventilation and vacuum pump guarantees fast cycles from 20 1 * 1 items
Catalog # CERT8500486
Supplier:  CERTOCLAV