Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Ochratoxin A standard solution at 1000 ng/ml in methanol 1 * 3 mL
RBIORBRP11A
:
R BIOPHARM
Ochratoxin A standard solution at 1000 ng/ml in methanol 1 * 3 mL
RBIORBRP11A
:
R BIOPHARM