Order Entry
Germany
Orders LinkContactUsLinkComponent
Polysorbate 80, J.T.Baker®

Polysorbate 80, J.T.Baker®

Jerrican, poly pail, 19 L
Catalog # X257-01
Supplier:   Avantor
Polysorbate 80, J.T.Baker®
Catalog # X257-01
Supplier:  Avantor
CAS Number:  9005-65-6
Polysorbate 80, J.T.Baker®
Polysorbate 80, J.T.Baker®

Jerrican, poly pail, 19 L

CAS Number:  9005-65-6
Price
Inquire For Price
Log in for availability
1 * 19 L
Product Details & Documents

Polyoxyethylene (20) Sorbitan Monooleate

Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Storage Temperature:Ambient
  • EINECS:500-018-3

Documents

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Identification (by IR):
    Passes Test

Specifications

  • Pack type:
    Jerrican, poly pail
  • Pk:
    19 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.