ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOACY-738 DEMONSTRATES INHIBITORY ACTIVITY AGAINST RECOMBINANT HDAC6 WITH IC50 VALUES OF 1.7 NM, WITH RESPECTIVE AVERAGE SELECTIVITY OVER CLASS I HDACS BEING 100-FOLD.IC50 VALUE: 1.7 NM 1 * 50 mg
Catalog # MDEXHY-19327-50MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOACY-738 DEMONSTRATES INHIBITORY ACTIVITY AGAINST RECOMBINANT HDAC6 WITH IC50 VALUES OF 1.7 NM, WITH RESPECTIVE AVERAGE SELECTIVITY OVER CLASS I HDACS BEING 100-FOLD.IC50 VALUE: 1.7 NM 1 * 50 mg
Catalog # MDEXHY-19327-50MG
Supplier:  MEDCHEMEXPRESS