ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO ≥5MG/ML WATER CYCLOPAMINE(11-DEOXOJERVINE) IS A SPECIFIC HEDGEHOG (HH) SIGNALING PATHWAY ANTAGONIST OF SMOOTHENED (SMO) WITH IC50 OF 46 NM.IC50 VALUE: 46 NM [1]TARGET 1 * 5 mg
Catalog # MDEXHY-17024-5MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO ≥5MG/ML WATER CYCLOPAMINE(11-DEOXOJERVINE) IS A SPECIFIC HEDGEHOG (HH) SIGNALING PATHWAY ANTAGONIST OF SMOOTHENED (SMO) WITH IC50 OF 46 NM.IC50 VALUE: 46 NM [1]TARGET 1 * 5 mg
Catalog # MDEXHY-17024-5MG
Supplier:  MEDCHEMEXPRESS