ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO:50MG/MLAP1903 IS A HOMODIMER BINDING TO FKBP ELICITES POTENT AND DOSEDEPE NDENT APOPTOTIC DEATH OF ENGINEERED CELL LINE HT1080 IN CULTURE WITH AN EC50 OF 0.1 NMIC50 VALUE: 0.1 NM (EC50, HT108 1 * 10 mg
Catalog # MDEXHY-16046-10MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO:50MG/MLAP1903 IS A HOMODIMER BINDING TO FKBP ELICITES POTENT AND DOSEDEPE NDENT APOPTOTIC DEATH OF ENGINEERED CELL LINE HT1080 IN CULTURE WITH AN EC50 OF 0.1 NMIC50 VALUE: 0.1 NM (EC50, HT108 1 * 10 mg
Catalog # MDEXHY-16046-10MG
Supplier:  MEDCHEMEXPRESS