ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: ≥ 30 MG/MLRO3280 IS A POTENT, HIGHLY SELECTIVE INHIBITOR OF POLO-LIKE KIN ASE 1 (PLK1) WITH IC50 OF 3 NM.IC50 VALUE: 3 NM (PLK1) [1]TARGET: PLKIN VITRO: DIFLUORO-ANALOG 7 (RO3280) GAVE THE BE 1 * 5 mg
Catalog # MDEXHY-15161-5MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: ≥ 30 MG/MLRO3280 IS A POTENT, HIGHLY SELECTIVE INHIBITOR OF POLO-LIKE KIN ASE 1 (PLK1) WITH IC50 OF 3 NM.IC50 VALUE: 3 NM (PLK1) [1]TARGET: PLKIN VITRO: DIFLUORO-ANALOG 7 (RO3280) GAVE THE BE 1 * 5 mg
Catalog # MDEXHY-15161-5MG
Supplier:  MEDCHEMEXPRESS