ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: >15 MG/MLTAK-715 IS A P38 MAPK INHIBITOR FOR P38Α WITH IC50 OF 7.1 NM, 28 -FOLD MORE SELECTIVE FOR P38Α OVER P38Β, NO INHIBITION TO P38Γ/Δ, JNK1, ERK1, IKKΒ, MEKK1 OR TAK1.IC50 VALUE: 7.1 NM 1 * 50 mg
Catalog # MDEXHY-10456-50MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: >15 MG/MLTAK-715 IS A P38 MAPK INHIBITOR FOR P38Α WITH IC50 OF 7.1 NM, 28 -FOLD MORE SELECTIVE FOR P38Α OVER P38Β, NO INHIBITION TO P38Γ/Δ, JNK1, ERK1, IKKΒ, MEKK1 OR TAK1.IC50 VALUE: 7.1 NM 1 * 50 mg
Catalog # MDEXHY-10456-50MG
Supplier:  MEDCHEMEXPRESS