ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: > 5.1 MG/ML, H2O: < 1 MG/MLCHIR-99021 (CT99021) IS A GSK-3Α/Β INHIBITOR W ITH IC50 OF 10 NM/6.7 NM > 500-FOLD SELECTIVITY FOR GSK-3 VERSUS ITS CLOSEST HOMOLOGS CDC2 AND ERK2, AS WELL AS OTHE 1 * 100 mg
Catalog # MDEXHY-10182-100MG
Supplier:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO: > 5.1 MG/ML, H2O: < 1 MG/MLCHIR-99021 (CT99021) IS A GSK-3Α/Β INHIBITOR W ITH IC50 OF 10 NM/6.7 NM > 500-FOLD SELECTIVITY FOR GSK-3 VERSUS ITS CLOSEST HOMOLOGS CDC2 AND ERK2, AS WELL AS OTHE 1 * 100 mg
Catalog # MDEXHY-10182-100MG
Supplier:  MEDCHEMEXPRESS