Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SOLVENT BLAU 4, REINHEIT: 95 % +
Catalog # XXXX86-55006-5G
Supplier:
Chemicals
Expand image
SOLVENT BLAU 4, REINHEIT: 95 % +
Catalog # XXXX86-55006-5G
Supplier:
Chemicals