ContactUsLinkComponent
Apc/fire™ 750 armenian hamster igg isotype ctrl. apc/fire™ 750 provides a more temperature and photo-stable alternative to apc/cy7. apc/fire™ 750 also has lower compensation requirements than apc/cy7 conjugates while maintaining an equal level of br 1 * 100 µG
Catalog # BLEG400962
Supplier:  BIOLEGEND INC
Apc/fire™ 750 armenian hamster igg isotype ctrl. apc/fire™ 750 provides a more temperature and photo-stable alternative to apc/cy7. apc/fire™ 750 also has lower compensation requirements than apc/cy7 conjugates while maintaining an equal level of br 1 * 100 µG
Catalog # BLEG400962
Supplier:  BIOLEGEND INC