ContactUsLinkComponent
Apc/fire™ 750 rat igg2a, kappa isotype ctrl. apc/fire™ 750 provides a more temperature and photo-stable alternative to apc/cy7. apc /fire™ 750 also has lower compensation requirements than apc/cy7 conjugates while maintaining an equal level of bright 1 * 100 µG
Catalog # BLEG400568
Supplier:  BIOLEGEND INC
Apc/fire™ 750 rat igg2a, kappa isotype ctrl. apc/fire™ 750 provides a more temperature and photo-stable alternative to apc/cy7. apc /fire™ 750 also has lower compensation requirements than apc/cy7 conjugates while maintaining an equal level of bright 1 * 100 µG
Catalog # BLEG400568
Supplier:  BIOLEGEND INC