ContactUsLinkComponent
Apc/fire 750 mouse igg2b, kappa isotype ctrl. apc/fire 750 provides a more temperature and photo-stable alternative to apc/cy7. a pc/fire 750 also has lower compensation requirements than apc/cy7 conjugates while maintaining an equal level of brig 1 * 100 Tests
Catalog # BLEG400372
Supplier:  BIOLEGEND INC
Apc/fire 750 mouse igg2b, kappa isotype ctrl. apc/fire 750 provides a more temperature and photo-stable alternative to apc/cy7. a pc/fire 750 also has lower compensation requirements than apc/cy7 conjugates while maintaining an equal level of brig 1 * 100 Tests
Catalog # BLEG400372
Supplier:  BIOLEGEND INC