ContactUsLinkComponent
Apc/fire™ 750 mouse igg2a, kappa isotype ctrl. apc/fire™ 750 provides a more temperature and photo-stable alternative to apc/cy7. a pc/fire™ 750 also has lower compensation requirements than apc/cy7 conjugates while maintaining an equal level of brig 1 * 25 Tests
Catalog # BLEG400283
Supplier:  BIOLEGEND INC
Apc/fire™ 750 mouse igg2a, kappa isotype ctrl. apc/fire™ 750 provides a more temperature and photo-stable alternative to apc/cy7. a pc/fire™ 750 also has lower compensation requirements than apc/cy7 conjugates while maintaining an equal level of brig 1 * 25 Tests
Catalog # BLEG400283
Supplier:  BIOLEGEND INC