ContactUsLinkComponent
Each vial of veri-cells pbmc contains 5 x 106 lyophilized human peripheral blood mononuclear cells (pbmc). 1 * 100 Tests
Catalog # BLEG425002
Supplier:  BIOLEGEND INC
Each vial of veri-cells pbmc contains 5 x 106 lyophilized human peripheral blood mononuclear cells (pbmc). 1 * 100 Tests
Catalog # BLEG425002
Supplier:  BIOLEGEND INC