ContactUsLinkComponent
True-nuclear™ one step staining human treg flow™ kit (foxp3 alexa fluor® 488/cd25 pe/cd4 percp) 1 * 25 Tests
Catalog # BLEG320127
Supplier:  BIOLEGEND INC
True-nuclear™ one step staining human treg flow™ kit (foxp3 alexa fluor® 488/cd25 pe/cd4 percp) 1 * 25 Tests
Catalog # BLEG320127
Supplier:  BIOLEGEND INC