ContactUsLinkComponent
Legendplex™ mouse cxcl13 (blc) capture bead b6, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles o f sandwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740101
Supplier:  BIOLEGEND INC
Legendplex™ mouse cxcl13 (blc) capture bead b6, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles o f sandwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740101
Supplier:  BIOLEGEND INC