ContactUsLinkComponent
Legendplex™ mouse ccl20 (mip-3alpha) capture bead a5, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the princi ples of sandwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740095
Supplier:  BIOLEGEND INC
Legendplex™ mouse ccl20 (mip-3alpha) capture bead a5, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the princi ples of sandwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740095
Supplier:  BIOLEGEND INC