ContactUsLinkComponent
Legendplex™ mouse ccl4 (mip-1beta) capture bead b5, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principl es of sandwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740091
Lieferant:  BIOLEGEND INC
Legendplex™ mouse ccl4 (mip-1beta) capture bead b5, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principl es of sandwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740091
Lieferant:  BIOLEGEND INC