ContactUsLinkComponent
Legendplex™ human cxcl5 (ena-78) capture bead b4, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740084
Supplier:  BIOLEGEND INC
Legendplex™ human cxcl5 (ena-78) capture bead b4, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740084
Supplier:  BIOLEGEND INC