ContactUsLinkComponent
Legendplex™ mouse il-17a capture bead b4, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sand wich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740061
Supplier:  BIOLEGEND INC
Legendplex™ mouse il-17a capture bead b4, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sand wich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740061
Supplier:  BIOLEGEND INC