ContactUsLinkComponent
Legendplex™ mouse il-10 capture bead b2, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandw ich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740059
Supplier:  BIOLEGEND INC
Legendplex™ mouse il-10 capture bead b2, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandw ich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740059
Supplier:  BIOLEGEND INC