ContactUsLinkComponent
Legendplex™ mouse il-5 capture bead a5, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandwi ch elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740056
Supplier:  BIOLEGEND INC
Legendplex™ mouse il-5 capture bead a5, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandwi ch elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740056
Supplier:  BIOLEGEND INC