ContactUsLinkComponent
Legendplex™ mouse il-4 capture bead a10, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandw ich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740055
Supplier:  BIOLEGEND INC
Legendplex™ mouse il-4 capture bead a10, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandw ich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740055
Supplier:  BIOLEGEND INC