ContactUsLinkComponent
Legendplex™ human tnf-alpha capture bead b3, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of s andwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740053
Supplier:  BIOLEGEND INC
Legendplex™ human tnf-alpha capture bead b3, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of s andwich elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740053
Supplier:  BIOLEGEND INC