ContactUsLinkComponent
Legendplex™ human il-5 capture bead a4, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandwi ch elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740043
Supplier:  BIOLEGEND INC
Legendplex™ human il-5 capture bead a4, 13x. biolegend’s legendplex™ is a bead-based immunoassay that uses the principles of sandwi ch elisas to quantify soluble analytes using a flow cytometer. 1 * 100 Tests
Catalog # BLEG740043
Supplier:  BIOLEGEND INC