Refractive Index 20C 1.44468 Solvent Ba 1 * 15 mL
Catalog # REGERI0144
Supplier: REAGECON
Frequently Bought Together
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.001 % H₂O)
Supplier: Merck
Acetone, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether, EMPARTA® ACS for analysis, Supelco®
Supplier: Merck
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific
1,4-Dioxane 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
n-Heptane, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Paraffin Oil heavy, liquid, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99%
Supplier: Thermo Fisher Scientific