Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ICP-METALLSTANDARDS MIT 14 KOMPONENTEN GE, NB, PD, PT, RH, SB, SN, TA, TI, W, ZR, IR, OS, RU MATRIX: 5% HNO3, 0,5% HCL, 0,5% HF, KONZENTRATION: JEWEILS 10 MG/L,
Catalog # EVOK7BE9105N05C05F
Supplier:
EVONIK MS PROJECT
Expand image
ICP-METALLSTANDARDS MIT 14 KOMPONENTEN GE, NB, PD, PT, RH, SB, SN, TA, TI, W, ZR, IR, OS, RU MATRIX: 5% HNO3, 0,5% HCL, 0,5% HF, KONZENTRATION: JEWEILS 10 MG/L,
Catalog # EVOK7BE9105N05C05F
Supplier:
EVONIK MS PROJECT