Orders LinkContactUsLinkComponent
Grounding mat and cables
Grounding mat and cables
Grounding cable, 1× clamp, 1x button plug, 1,5 m
Catalog # SCTV108222
Supplier:   S.C.A.T.
Grounding mat and cables
Catalog # SCTV108222
Supplier:  S.C.A.T.
Grounding mat and cables
Grounding mat and cables
Grounding cable, 1× clamp, 1x button plug, 1,5 m
Price
40,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
In order to prevent static discharge with fluids in plastic containers, the contents of the waste receptacles can be secured with ground connection. This allows users to establish an additional safety factor when collecting combustible fluids.
  • Grounding cable with different connection types
  • Cable lengths from 1,5 to 3 metres available
  • Matching with Grounding plug EU 1 megohm for connection to building services
With the different grounding cables users have the possibility to safely conduct the electrostatic charge and prevent the risk of explosion due to flying sparks. The grounding cables are suitable for all SCAT and SymLine products, as well as most other suppliers.
Specifications
  • Description:
    Grounding cable, 1× clamp, 1x button plug
  • Length:
    1,5 m
  • Seq:
    08