ContactUsLinkComponent
ZOOM BINOCULAR HEAD, WITH EYEP. 10:1 ZOOM RATIO 1 * 1 items
Catalog # OPTKSZP-10
Supplier:  OPTIKA
ZOOM BINOCULAR HEAD, WITH EYEP. 10:1 ZOOM RATIO 1 * 1 items
Catalog # OPTKSZP-10
Supplier:  OPTIKA