Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Safety-can 1/2l with screw cap 1inch, flame trap and pressure control valve - unpolished 1 * 1 items
Catalog # ROET0HKU
Supplier:
ROETZMEIER SICHERH BEHAELTER
Expand image
Safety-can 1/2l with screw cap 1inch, flame trap and pressure control valve - unpolished 1 * 1 items
Catalog # ROET0HKU
Supplier:
ROETZMEIER SICHERH BEHAELTER