Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MacConkey Agar w/1.35% Agar, CV, NaCl and 0.15% Bile Salts 1 * 500 g
Catalog # HIMEM081D-500G
Supplier:
HIMEDIA
Expand image
MacConkey Agar w/1.35% Agar, CV, NaCl and 0.15% Bile Salts 1 * 500 g
Catalog # HIMEM081D-500G
Supplier:
HIMEDIA