Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant proteins. This product is prepared by labeling high quality monoclonal mouse anti-GFP IgG (H+L) with the NIR fluorescent dye CF™680R. 1 * 100 µl
Monoclonal anti-GFP is designed to detect enhanced GFP and GFP containing recombinant proteins. This product is prepared by labeling high quality monoclonal mouse anti-GFP IgG (H+L) with the NIR fluorescent dye CF™680R. 1 * 100 µl