Tray Quatrz glass in application temperatures of up to 1050°C (maximum 1600°C), 3-sided, with open end, translucent, high chemical resistance and purity. Length 248mm, width 152mm, height 35mm. The durability or operating cycles of our quartz glass n 1 * 1 items
Catalog # TSLA73043
Supplier: TECHNICAL QUARTZ SOLUTIONS