Orders LinkContactUsLinkComponent
Covernets
Covernets
Covering net, 215×445 mm
Catalog # MIEL10283650
Supplier:   MIELE
Covernets
Catalog # MIEL10283650
Supplier:  MIELE
Covernets
Covernets
Covering net, 215×445 mm
Price
165,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Covering net for optimum fixing of delicate items to be washed.
  • Stainless steel frame covered with plastic net
  • Particularly resistant and durable
Specifications
  • Description:
    Covering net, 215×445 mm
  • Seq:
    01