Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Color Standard, APHA/Hazen (Platinum- Cobalt), color = 30 1 * 500 mL
Catalog # RICCR2230030-500C
Supplier:
Ricca Chemical
Expand image
Color Standard, APHA/Hazen (Platinum- Cobalt), color = 30 1 * 500 mL
Catalog # RICCR2230030-500C
Supplier:
Ricca Chemical