VESPEL FERRULE GVF16-004 1 * 10 items
Catalog # SHIM670-15003-03
Supplier: SHIMADZU
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Latex Gloves
Supplier: VWR Collection