Order Entry
Germany
Orders LinkContactUsLinkComponent
CULTREX BME TYPE 2 PATHCLEAR 1 * 5 ml

CULTREX BME TYPE 2 PATHCLEAR 1 * 5 ml

Catalog # TREV3532-005-02
Marketsource
Supplier:   TREVIGEN
CULTREX BME TYPE 2 PATHCLEAR 1 * 5 ml
Catalog # TREV3532-005-02
Supplier:  TREVIGEN
Marketsource
CULTREX BME TYPE 2 PATHCLEAR 1 * 5 ml
Price
Inquire For Price
Log in for availability
1 * 5 mL
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.