Orders LinkContactUsLinkComponent
Drape sheets, Foliodrape® Protect
Drape sheets, Foliodrape® Protect
Foliodrape® Protect drape sheets, sterile, in 4-layer packaging, self-adhesive, 750×900 mm
Catalog # HART2775082
Supplier:   HARTMANN PAUL
Drape sheets, Foliodrape® Protect
Catalog # HART2775082
Supplier:  HARTMANN PAUL
Drape sheets, Foliodrape® Protect
Drape sheets, Foliodrape® Protect
Foliodrape® Protect drape sheets, sterile, in 4-layer packaging, self-adhesive, 750×900 mm
Price
97,00 €
Log in for availability
1 * 40 ST
Product Details & Documents
These drape sheets are made from a soft polyethylene foil laminated to an absorbent non woven top layer. For sterile draping of patients and equipment during operations with moderate quantities of fluids, especially in outpatient settings.
  • Impermeable to moisture and bacteria
  • Abrasion resistant and extremely low linting
  • Highly conformable
  • Also available as an adhesive drape with self-adhesive area for rapid and accurate positioning at the incision site
Certifications: DIN 13795 (1-3)
Specifications
  • Description:
    Foliodrape® Protect drape sheets, sterile, in 4-layer packaging, self-adhesive
  • L×W:
    750×900 mm