The Snail family of developmental regulatory proteins is a group of widely conserved zinc-finger proteins that regulate transcriptio n and include the mammalian proteins SLUG, SNAI 1, the human homolog of Drosophila SNAIL, and Smuc. 1 * 100 µG
Catalog # BWRLBS8479
Supplier: Bioworld Technology