Columns, PlusPore
Analytical, 2,10 mm, 250 mm
Catalog # HEWLPL1913-5520
Supplier: Agilent
Jump to:
Product Details & Documents
The PlusPore family offers a major resolution improvement over mixed medias and wider range than single pore columns.
- Separation is maximised over four common MW ranges
- Small particles and large pore volumes give very fast, high resolution
- High speed chromatography offers 6 min separations for common samples
The presence of multiple pore sizes on each particle maximises the separation efficiency while still offering a wide MW range with no dislocations.
Specifications
- Column Format:Analytical
- Ø int.:2,10 mm
- Length:250 mm
- Seq:04



