Orders LinkContactUsLinkComponent
Spray chambers and brackets
Spray chambers and brackets
Spray chamber, single-pass, for 5100 ICP-OES, glass cyclonic design with ball joint socket and UniFit drain outlet, for aqueous and low matrix samples where best sensitivity and lowest LODs are required
Catalog # HEWLG8010-60271
Supplier:   Agilent
Spray chambers and brackets
Catalog # HEWLG8010-60271
Supplier:  Agilent
Spray chambers and brackets
Spray chambers and brackets
Spray chamber, single-pass, for 5100 ICP-OES, glass cyclonic design with ball joint socket and UniFit drain outlet, for aqueous and low matrix samples where best sensitivity and lowest LODs are required
Price
875,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Specifications
  • Description:
    Spray chamber, single-pass, for 5100 ICP-OES, glass cyclonic design with ball joint socket and UniFit drain outlet, for aqueous and low matrix samples where best sensitivity and lowest LODs are required
  • Seq:
    05